Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LPPR2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561851
Description
Immunogen sequence: VHNFQSRPPS GRRLSPWEDL GQAPTMDSPL EKNPRSAGRI RHRHGSPHPS RRTA Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.
LPPR2 is a multi-pass membrane protein and belongs to the PA-phosphatase related phosphoesterase family. LPPR2 shows dynamic expression regulation during brain development and neuronal excitation.
Specifications
LPPR2 | |
Polyclonal | |
Unconjugated | |
PLPPR2 | |
Lipid phosphate phosphatase-related protein type 2; Lppr2; MGC137198 protein; phospholipid phosphatase related 2; phospholipid phosphatase-related protein type 2; plasticity related gene 4; plasticity related protein 4; plasticity-related gene 4 protein; PLPPR2; PRG4; PRG-4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64748 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96GM1 | |
PLPPR2 | |
Recombinant protein corresponding to Human LPPR2. Recombinant protein control fragment (Product #RP-101177). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction