Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LPPR2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$219.50 - $487.50
Specifications
Antigen | LPPR2 |
---|---|
Dilution | Western Blot, ELISA |
Applications | Western Blot, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
LPPR2 Polyclonal antibody specifically detects LPPR2 in Human samples. It is validated for Western Blot, ELISASpecifications
LPPR2 | |
Western Blot, ELISA | |
Unconjugated | |
Rabbit | |
Human | |
DKFZp761E1121, EC 3.1.3.4, FLJ13055, lipid phosphate phosphatase-related protein type 2, plasticity related gene 4, Plasticity-related gene 4 protein, PRG4, PRG-4 | |
Synthetic peptide derived from the LPPR2 protein (Uniprot: Q96GM1) | |
Primary | |
Store at -70C. Avoid freeze-thaw cycles. |
Western Blot, ELISA | |
Polyclonal | |
Purified | |
RUO | |
PBS | |
64748 | |
IgG | |
Antigen Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title