Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LPPR3 Polyclonal Antibody

Catalog No. PIPA563390
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563390 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563390 Supplier Invitrogen™ Supplier No. PA563390

Rabbit Polyclonal Antibody

Immunogen sequence: VGNFQAPPAE KPAAPAPAKD ALRALTQRGH DSVYQQNKSV STDEL Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 87%.

Phospholipid-mediated signalling on neurons provokes diverse responses such as neurogenesis, pattern formation and neurite remodelling. A recently uncovered novel set of molecules in the mammalian brain, named plasticity-related genes (PRGs) mediate lipid phosphate phosphatase activity and provide evidence for their involvement in mechanisms of neuronal plasticity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LPPR3
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PLPPR3
Gene Accession No. Q6T4P5
Gene Alias BMPG; Bone marrow proteoglycan; bone-marrow proteoglycan; EMBP; Eosinophil granule major basic protein; eosinophil major basic protein; lipid phosphate phosphatase-related protein type 3; LPPR3; LPR3; MBP; MBP1; natural killer cell activator; PAP-2-like protein 2; phospholipid phosphatase related 3; phospholipid phosphatase-related protein type 3; PHP2; Plasticity-related gene 2 protein; PLPPR3; Pregnancy-associated major basic protein; PRG2; PRG-2; proMBP; Proteoglycan 2; proteoglycan 2 preproprotein; proteoglycan 2, bone marrow (natural killer cell activator, eosinophil granule major basic protein); proteoglycan 2, pro eosinophil major basic protein
Gene Symbols PLPPR3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LPPR3.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79948
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.