Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LPPR4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA580908
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA580908 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA580908 Supplier Invitrogen™ Supplier No. PA580908
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human LPPR4.

Phospholipid-mediated signalling on neurons provokes diverse responses such as neurogenesis, pattern formation and neurite remodelling. A recently uncovered novel set of molecules in the mammalian brain, named plasticity-related genes (PRGs) mediate lipid phosphate phosphatase activity and provide evidence for their involvement in mechanisms of neuronal plasticity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LPPR4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene PLPPR4
Gene Accession No. Q7Z2D5
Gene Alias A330086D10; Brain-specific phosphatidic acid phosphatase-like protein 1; D3Bwg0562e; Kiaa0455; Lipid phosphate phosphatase-related protein type 4; LPPR4; LPR4; mKIAA0455; phospholipid phosphatase related 4; phospholipid phosphatase-related protein type 4; Php1; plasticity related gene 1; plasticity related gene-1; plasticity-related gene 1 protein; PLPPR4; Prg1; Prg-1; RP4-788L13.1
Gene Symbols PLPPR4
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the center region of the Human LPPR4.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9890
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.