Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LPPR4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01674855
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-674-855 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-674-855 Supplier Invitrogen™ Supplier No. PA112728
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control for this product is human brain lysate.

Phospholipid-mediated signalling on neurons provokes diverse responses such as neurogenesis, pattern formation and neurite remodelling. A recently uncovered novel set of molecules in the mammalian brain, named plasticity-related genes (PRGs) mediate lipid phosphate phosphatase activity and provide evidence for their involvement in mechanisms of neuronal plasticity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LPPR4
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.08% sodium azide
Gene PLPPR4
Gene Accession No. Q7Z2D5
Gene Alias A330086D10; Brain-specific phosphatidic acid phosphatase-like protein 1; D3Bwg0562e; Kiaa0455; Lipid phosphate phosphatase-related protein type 4; LPPR4; LPR4; mKIAA0455; phospholipid phosphatase related 4; phospholipid phosphatase-related protein type 4; Php1; plasticity related gene 1; plasticity related gene-1; plasticity-related gene 1 protein; PLPPR4; Prg1; Prg-1; RP4-788L13.1
Gene Symbols PLPPR4
Host Species Rabbit
Immunogen Synthetic peptide corresponding to LPPRP4.
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9890
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.