Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LPPR4 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981744
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1744 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1744 Supplier Bioss Supplier No. BS11875R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Phospholipid-mediated signalling on neurons provokes diverse responses such as neurogenesis, pattern formation and neurite remodelling. A recently uncovered novel set of molecules in the mammalian brain, named plasticity-related genes (PRGs) mediate lipid phosphate phosphatase activity and provide evidence for their involvement in mechanisms of neuronal plasticity.

Specifications

Antigen LPPR4
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PLPPR4
Gene Accession No. Q7Z2D5
Gene Alias Brain-specific phosphatidic acid phosphatase-like protein 1; KIAA0455; Lipid phosphate phosphatase-related protein type 4; LPPR4; LPR4; phospholipid phosphatase related 4; Phospholipid phosphatase-related protein type 4; PHP1; plasticity related gene 1; plasticity related gene-1; Plasticity-related gene 1 protein; PLPPR4; PRG1; PRG-1; RP4-788L13.1
Gene Symbols PLPPR4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human LPPR4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9890
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.