Learn More
Invitrogen™ LPPR5 Polyclonal Antibody

Description
The antibody detects endogenous levels of total PAP2D / LPPR5 protein.
Specifications
Specifications
| Antigen | LPPR5 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 2.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | PLPPR5 |
| Gene Accession No. | Q32ZL2, Q8BJ52 |
| Gene Alias | 4833424O15Rik; Lipid phosphate phosphatase-related protein type 5; Lppr5; PAP2; Pap2d; phosphatidic acid phosphatase 2d; phosphatidic acid phosphatase type 2; phosphatidic acid phosphatase type 2d; phospholipid phosphatase related 5; phospholipid phosphatase-related protein type 5; Plasticity-related gene 5 protein; plasticity-related protein 5; PLPPR5; PRG5; PRG-5 |
| Gene Symbols | PLPPR5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.