Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ lprG Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144398
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144398 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144398 Supplier Invitrogen™ Supplier No. PA5144398
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

lprG Polyclonal Antibody for Western Blot, ELISA

Probably helps membrane protein MT1454 (P55) transport triacylglycerides (TAG) across the inner cell membrane into the periplasm and probably ultimately to the outer membrane. TAG probably regulates lipid metabolism and growth regulation. Binds di- and triacylated phosphatidyl-myo-inositol mannosides (PIMs), and glycolipid lipoglycan modulins lipoarabinomannan (LAM) and lipomannan (LM), facilitating their recognition by TLR2. Required for activity of drug efflux transporter MT1454. Required, probably with MT1454, for normal surface localization of LAM.
TRUSTED_SUSTAINABILITY

Specifications

Antigen lprG
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.977 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene lprG
Gene Accession No. P9WK44
Gene Alias lpp-27
Gene Symbols lprG
Host Species Rabbit
Immunogen Recombinant Mycobacterium tuberculosis lprG protein (27-236aa ).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 886700
Target Species Bacteria
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.