Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LPXN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560491
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560491 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560491 Supplier Invitrogen™ Supplier No. PA560491
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AQLVYTTNIQ ELNVYSEAQE PKESPPPSKT SAAAQLDELM AHLTEMQAKV AVRADAGKKH LPDKQDHKAS LDSML Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 81%.

The product encoded by this gene is preferentially expressed in hematopoietic cells and belongs to the paxillin protein family. Similar to other members of this focal-adhesion-associated adaptor-protein family, it has four leucine-rich LD-motifs in the N-terminus and four LIM domains in the C-terminus. It may function in cell type-specific signaling by associating with PYK2, a member of focal adhesion kinase family. As a substrate for a tyrosine kinase in lymphoid cells, this protein may also function in, and be regulated by, tyrosine kinase activity. Alternative splicing results in multiple transcript variants encoding distinct isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LPXN
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LPXN
Gene Accession No. O60711
Gene Alias 4933402K05Rik; A530083L21Rik; AV278559; LDLP; LDPL; Leupaxin; LPXN
Gene Symbols LPXN
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LPXN. Recombinant protein control fragment (Product #RP-100453).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9404
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.