Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LPXN Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA597933
Description
LPXN Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
The product encoded by this gene is preferentially expressed in hematopoietic cells and belongs to the paxillin protein family. Similar to other members of this focal-adhesion-associated adaptor-protein family, it has four leucine-rich LD-motifs in the N-terminus and four LIM domains in the C-terminus. It may function in cell type-specific signaling by associating with PYK2, a member of focal adhesion kinase family. As a substrate for a tyrosine kinase in lymphoid cells, this protein may also function in, and be regulated by, tyrosine kinase activity. Alternative splicing results in multiple transcript variants encoding distinct isoforms.
Specifications
LPXN | |
Polyclonal | |
Unconjugated | |
LPXN | |
4933402K05Rik; A530083L21Rik; AV278559; LDLP; LDPL; Leupaxin; LPXN | |
Rabbit | |
Protein G | |
RUO | |
9404 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
3 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
O60711 | |
LPXN | |
Recombinant Human Leupaxin protein (1-300AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction