Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LR8 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | LR8 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
LR8 Polyclonal specifically detects LR8 in Human samples. It is validated for Western Blot.Specifications
LR8 | |
Polyclonal | |
Rabbit | |
NP_054739 | |
28959 | |
Synthetic peptide directed towards the C terminal of human TMEM176B. Peptide sequence SLVSLGVGLRNLCGQSSQPLNEEGSEKRLLGENSVPPSPSREQTSTAIVL. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
LR8FLJ26014, LR8-like protein, MGC110857, Protein LR8, transmembrane protein 176B | |
TMEM176B | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title