Learn More
LRAT Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA593123
Description
Immunogen sequence: MKNPMLEVVS LLLEKLLLIS NFTLFSSGAA GEDKGRNSFY ETSSFHRGDV LEVPRTHLTH YGIYLGDNRV AHMMPDILLA LTDDMGRTQK VVSNKRLILG VIVKVASIRV DTVEDFAYGA NILVNHLDES LQKKALLNEE VARRAEKLLG FTPYSLLWNN CEHFVTYCRY GTPISPQSDK FCETVKIIIR DQRS; Positive Samples: Rat testis; Cellular Location: Cytoplasm, Endoplasmic reticulum membrane, Endosome, Rough endoplasmic reticulum, Single-pass membrane protein, multivesicular body, perinuclear region
Lecithin retinol acyltransferase (LRAT) is a membrane bound enzyme that catalyzes the transfer of an acyl group from the sn-1 position of lecithin to vitamin A which generates all-trans-retinyl esters (tREs) in the liver, some extrahepatic tissues, such as the lung, and retinal pigmented epithelium. LRAT can also exchange palmitoyl groups between RPE65, a tRE binding protein essential for vision, and tREs, which is important for the operation of the visual pathway. LRAT is essential for the dietary mobilization, transport, and storage of vitamin A as well as the synthesis of the visual pigment chromophore. LRAT monomers interact in membranes to form homodimers through disulfide bond formation. A loss of LRAT correlates with an early onset severe retinal dystrophy and severe retinyl ester deprivation, while a reduction in LRAT expression may be associated with invasive bladder cancer.
Specifications
LRAT | |
Polyclonal | |
Unconjugated | |
LRAT | |
1300010A18Rik, AI449251, LCA14, Lecithin retinol acyltransferase, lecithin retinol acyltransferase (phosphatidylcholine--retinol O-acyltransferase), lecithin-retinol acyltransferase (phosphatidylcholine-retinol-O-acyltransferase), Lrat, Phosphatidylcholine--retinol O-acyltransferase, phosphatidylcholine-retinol-O-acyltransferase | |
Rabbit | |
Affinity Chromatography | |
RUO | |
64047, 9227 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Western Blot | |
0.34 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide, pH 7.3 | |
O95237, Q9JI61 | |
LRAT | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-194 of human LRAT (NP_0047352) | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.