Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRBA Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$382.00 - $610.00
Specifications
Antigen | LRBA |
---|---|
Dilution | Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:200-1:500, Knockdown Validated |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
LRBA Polyclonal specifically detects LRBA in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
LRBA | |
Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
CDC4L, CDC4-like protein, DKFZp686A09128, DKFZp686K03100, FLJ16600, LAB300, LBA, lipopolysaccharide-responsive and beige-like anchor protein, LPS-responsive vesicle trafficking, beach and anchor containing, MGC72098, vesicle trafficking, beach and anchor containing | |
LRBA | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:200-1:500, Knockdown Validated | |
Polyclonal | |
Rabbit | |
Human | |
P50851 | |
987 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:GGWRVWVDTLSITHSKVTFEIHKENLANIFREQQGKVDEEIGLCSSTSVQAASGIRRDINVSVGSQQPDTKDSPVCPHFTTNGNENSSIEKTSSLESASNIELQTTNTSYEEMKAEQENQELPDEGTLEETL | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title