Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRBA Mouse anti-Human, Clone: 2D4.1, MilliporeSigma™

Catalog No. MABF109625 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF1096-25 25 μg
MABF1096-100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF1096-25 Supplier MilliporeSigma Supplier No. MABF109625UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-LRBA, clone 2D4.1 Antibody.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRBA
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2D4.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols LRBA,BGL,CDC4L,LBA
Host Species Mouse
Immunogen GST/His-tagged recombinant fragment corresponding to 115 amino acids from the N-terminal half of human Lipopolysaccharide-responsive and beige-like anchor protein (LRBA).
Purification Method Protein G Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_006717
Test Specificity Clone 2D4.1 detects human Lipopolysaccharide-responsive and beige-like anchor protein (LRBA). It targets an epitope within 115 amino acids from the N-terminal half.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.