Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LRBA Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553986
Description
Immunogen sequence: SVLMVSKYRD ILEPQNERHS QSCTETGSEN ENVSLSEITP AAFSTLTTAS VEESESTSSA RRRDSGIGEE TATGLGSHVE VTPHTAPPGV SAGPDAISEV LSTLSLEVNK SPETKNDRGN DLDTKATPSV SV Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 73%.
The protein encoded by this gene is a member of the WDL-BEACH-WD (WBW) gene family. Its expression is induced in B cells and macrophages by bacterial lipopolysaccharides (LPS). The encoded protein associates with protein kinase A and may be involved in leading intracellular vesicles to activated receptor complexes, which aids in the secretion and/or membrane deposition of immune effector molecules. Defects in this gene are associated with the disorder common variable immunodeficiency-8 with autoimmunity. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
LRBA | |
Polyclonal | |
Unconjugated | |
LRBA | |
Beige-like protein; BGL; CDC4L; CDC4-like protein; CVID8; LAB300; LBA; Lipopolysaccharide-responsive and beige-like anchor protein; LPS responsive beige-like anchor protein; LPS-responsive vesicle trafficking, beach and anchor containing; LRBA; vesicle trafficking, beach and anchor containing | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
987 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P50851 | |
LRBA | |
Recombinant protein corresponding to Human LRBA. Recombinant protein control fragment (Product #RP-92444). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction