Learn More
LRBA Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304478A

Description
The recommended shelf life for this product is 1 year from date of receipt.
The protein encoded by this gene is a member of the WDL-BEACH-WD (WBW) gene family. Its expression is induced in B cells and macrophages by bacterial lipopolysaccharides (LPS). The encoded protein associates with protein kinase A and may be involved in leading intracellular vesicles to activated receptor complexes, which aids in the secretion and/or membrane deposition of immune effector molecules. Defects in this gene are associated with the disorder common variable immunodeficiency-8 with autoimmunity. Two transcript variants encoding different isoforms have been found for this gene.Specifications
LRBA | |
Polyclonal | |
Unconjugated | |
LRBA | |
Beige-like protein, BGL, CDC4L, CDC4-like protein, CVID8, LAB300, LBA, LPS-responsive vesicle trafficking, beach and anchor containing, vesicle trafficking, beach and anchor containing | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
987 | |
4° C | |
Liquid |
Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
P50851 | |
LRBA | |
Between 1125 and 1175 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.