Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRC58 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA571147
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571147 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571147 Supplier Thermo Scientific Supplier No. PA571147

Rabbit Polyclonal Antibody

LRRC58 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRC58
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene LRRC58
Gene Accession No. Q96CX6
Gene Alias leucine rich repeat containing 58; leucine-rich repeat-containing protein 58; LRRC58
Gene Symbols LRRC58
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of human LRC58
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116064
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.