Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRCH1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP287737
Description
LRCH1 Polyclonal specifically detects LRCH1 in Human samples. It is validated for Western Blot.Specifications
LRCH1 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
23143 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
CHDC1, FLJ41660, KIAA1016, leucine-rich repeat and calponin homology domain-containing protein 1, leucine-rich repeats and calponin homology (CH) domain containing 1, neuronal protein 81, NP81 | |
The immunogen is a synthetic peptide directed towards the middle region of Human LRCH1 | |
100 μL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction