Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRCH2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5145148
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145148 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5145148 Supplier Invitrogen™ Supplier No. PA5145148
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RNKPKQTVEC EKSVSADEVN SPLSPLTWQP LENQKDQIDE QPWPESHPII WQSEERRRSK QIRKEYFKYK SMRKSSSGNE NDEQDSDNAN MSTQSPVS Antigen sequence identity: Mouse 85%; Rat 86%.

Leucine rich repeat and calponin homology domain containing protein 2 (LRCH2) also named as KIAA1495 is a 765 amino acid protein, which contains one CH domain and nine LRR repeats. The function of LRCH2 is still non-known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRCH2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LRCH2
Gene Accession No. Q5VUJ6
Gene Alias AI316885; dA204F4.4; Kiaa1495; leucine rich repeats and calponin homology domain containing 2; leucine-rich repeat and calponin homology domain-containing protein 2; leucine-rich repeats and calponin homology (CH) domain containing 2; Lrch2; mKIAA1495
Gene Symbols LRCH2
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 498-595. Recombinant protein control fragment (Product #RP-109954).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57631
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.