Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LRCH3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552933
Description
Immunogen sequence: HASPLPPSAA PTTDSTDSIT GQNSRQREEE LELIDQLRKH IEYRLKVSLP CDLGAALTDG VVLCHLANHV RPRSVPSIHV PSP Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 84%.
LRCH3 (Leucine Rich Repeats And Calponin Homology Domain Containing 3) is a Protein Coding gene.
Specifications
LRCH3 | |
Polyclonal | |
Unconjugated | |
Lrch3 | |
2210409B11Rik; AW215594; DISP complex protein LRCH3; Gm1742; leucine rich repeats and calponin homology domain containing 3; leucine-rich repeat and calponin homology domain-containing protein 3; leucine-rich repeats and calponin homology (CH) domain containing 3; LRCH3; MGC4126 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
70144, 84859 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.30 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8BVU0, Q96II8 | |
Lrch3 | |
Recombinant protein corresponding to Human LRCH3. Recombinant protein control fragment (Product #RP-90560). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction