Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRCH3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564970
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564970 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564970 Supplier Invitrogen™ Supplier No. PA564970
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NLCSPSDILQL NLSVKRTVET LLSLGAHSEE SSFVCLSL Highest antigen sequence identity to the following orthologs - mouse 87%, rat 38%.

LRCH3 (Leucine Rich Repeats And Calponin Homology Domain Containing 3) is a Protein Coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRCH3
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Lrch3
Gene Accession No. Q96II8
Gene Alias 2210409B11Rik; AW215594; DISP complex protein LRCH3; Gm1742; leucine rich repeats and calponin homology domain containing 3; leucine-rich repeat and calponin homology domain-containing protein 3; leucine-rich repeats and calponin homology (CH) domain containing 3; LRCH3; MGC4126
Gene Symbols Lrch3
Host Species Rabbit
Immunogen Recombinant Human LRCH3. Recombinant protein control fragment (Product #RP-105742).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84859
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.