Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRCH4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP169265
Description
LRCH4 Polyclonal specifically detects LRCH4 in Human samples. It is validated for Western Blot.Specifications
LRCH4 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
FLJ40101, FLJ46315, leucine rich repeat neuronal 4, Leucine-rich neuronal protein, leucine-rich repeat and calponin homology domain-containing protein 4, Leucine-rich repeat neuronal protein 4, leucine-rich repeats and calponin homology (CH) domain containing 4, LRN, LRRN1, LRRN4, PP14183 | |
Rabbit | |
73 kDa | |
100 μL | |
Primary | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
O75427 | |
LRCH4 | |
Synthetic peptides corresponding to LRCH4(leucine-rich repeats and calponin homology (CH) domain containing 4) The peptide sequence was selected from the middle region of LRCH4. Peptide sequence PGHRYDGGLDSGFHSVDSGSKRWSGNESTDEFSELSFRISELAREPRGPR The peptide sequence for this immunogen was taken from within the described region. | |
Affinity purified | |
RUO | |
4034 | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Yeast, Zebrafish | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction