Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRCH4 Monoclonal Antibody (OTI1F8), TrueMAB™, OriGene
SDP

Catalog No. 502539823
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-253-9823 30 μL
50-167-5766 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-253-9823 Supplier OriGene Supplier No. TA505246S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LRCH4 Monoclonal antibody specifically detects LRCH4 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen LRCH4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1F8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene PPIL3
Gene Accession No. Q9H2H8
Gene Alias 2810008P14Rik; 2900069C24Rik; AI558103; FLJ40101; FLJ46315; leucine rich repeat neuronal 4; leucine rich repeat protein 4, neuronal; leucine rich repeats and calponin homology domain containing 4; leucine-rich neuronal protein; Leucine-rich repeat and calponin homology domain-containing protein 4; Leucine-rich repeat neuronal protein 4; leucine-rich repeats and calponin homology (CH) domain containing 4; Lrch4; LRN; LRRN1; LRRN4; mFLJ00248; PP14183; SAP25
Host Species Mouse
Immunogen Full length human recombinant protein of human PPIL3 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53938
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.