Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRCH4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558049
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558049 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558049 Supplier Invitrogen™ Supplier No. PA558049
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LPIAGPATAP APRPLGSIQR PNSFLFRSSS QSGSGPSSPD SVLRPRRYPQ VPDEKDLMTQ LRQVLESRLQ Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 79%.

LRCH4 is a protein that contains leucine-rich repeats (LRR) at its amino terminus and that is known to be involved in ligand binding. The carboxyl terminus may act as a membrane anchor. Identified structural elements suggest that this protein resembles a receptor.This gene encodes a protein that contains leucine-rich repeats (LRR) at its amino terminus and that is known to be involved in ligand binding. The carboxyl terminus may act as a membrane anchor. Identified structural elements suggest that the encoded protein resembles a receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRCH4
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LRCH4
Gene Accession No. O75427
Gene Alias 2810008P14Rik; 2900069C24Rik; AI558103; FLJ40101; FLJ46315; leucine rich repeat neuronal 4; leucine rich repeat protein 4, neuronal; leucine rich repeats and calponin homology domain containing 4; leucine-rich neuronal protein; Leucine-rich repeat and calponin homology domain-containing protein 4; Leucine-rich repeat neuronal protein 4; leucine-rich repeats and calponin homology (CH) domain containing 4; Lrch4; LRN; LRRN1; LRRN4; mFLJ00248; PP14183; SAP25
Gene Symbols LRCH4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LRCH4. Recombinant protein control fragment (Product #RP-96542).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4034
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.