Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRFN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567434
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567434 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567434 Supplier Invitrogen™ Supplier No. PA567434
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PWRIPPSAPRP KPSLDRLMGA FASLDLKSQR KEELLDSRTP AGRGAGTSAR GHHSDREPLL Highest antigen sequence identity to the following orthologs - mouse 95%, rat 97%.

LRFN2 is one of a family of five transmembrane glycoproteins that are highly expressed in neuronal tissues. LRFN proteins share leucine-rich repeat (LRR)-immunoglobulin-like (Ig)-fibronectin type III (Fn)-transmembrane domain structure with other members of the LRR-Ig-Fn protein superfamily such as the Slitrk family of proteins. Expression of LRFN1, -3, and -4 mRNA was detected in embryonic neuronal cells, while Lrfn2 and Lrfn5 expression was primarily restricted to more mature cells. LRFN1, -2, and -4 bound to PDZ domains of postsynaptic PSD95, re-distributing PSD95 to the cell periphery. It has been suggested that the Lrfn proteins play a role in the developing and/or mature vertebrate nervous system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRFN2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LRFN2
Gene Accession No. Q9ULH4
Gene Alias 5730420O05Rik; fibronectin type III, immunoglobulin and leucine rich repeat domains 2; FIGLER2; KIAA1246; leucine rich repeat and fibronectin type III domain containing 2; leucine-rich repeat and fibronectin type-III domain-containing protein 2; Lrfn2; mKIAA1246; RP11-535K1.2; SALM1; synaptic adhesion-like molecule 1
Gene Symbols LRFN2
Host Species Rabbit
Immunogen Recombinant Human LRFN2. Recombinant protein control fragment (Product #RP-108095).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57497
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.