Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRFN3 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA520706

Catalog No. PIPA520706


Only null left
Add to Cart

Description

Description

A suggested positive control is K562 cell lysate. PA5-20706 can be used with blocking peptide PEP-0821.

LRFN3 is one of a family of five transmembrane glycoproteins that are highly expressed in neuronal tissues. LRFN proteins share leucine-rich repeat (LRR)-immunoglobulin-like (Ig)-fibronectin type III (Fn)-transmembrane domain structure with other members of the LRR-Ig-Fn protein superfamily such as the Slitrk family of proteins. Expression of LRFN1, -3, and -4 mRNA was detected in embryonic neuronal cells, while Lrfn2 and Lrfn5 expression was primarily restricted to more mature cells. LRFN1, -2, and -4 bound to PDZ domains of postsynaptic PSD95, re-distributing PSD95 to the cell periphery. It has been suggested that the Lrfn proteins play a role in the developing and/or mature vertebrate nervous system.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LRFN3
Polyclonal
Unconjugated
LRFN3
A530045B06Rik; EMBL:AAI58863.1, ECO:0000312; fibronectin type III, immunoglobulin and leucine rich repeat domains 1; FIGLER1; leucine rich repeat and fibronectin type III domain containing 3; leucine-rich repeat and fibronectin type-III domain-containing protein 3; LRFN3; lrfn3 {ECO:0000312; PubMed:18227064}; RGD:1305567}; Salm4; Synaptic adhesion-like molecule 4; synaptic adhesion-like molecule 4 {ECO:0000303; UNQ5865/PRO34192
Rabbit
Antigen affinity chromatography
RUO
233067, 308495, 79414
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Liquid
Immunohistochemistry, Western Blot
1 mg/mL
PBS with 0.02% sodium azide
B0BNK7, Q8BLY3, Q9BTN0
LRFN3
A 16 amino acid peptide near the center of human LRFN3.
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.