Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LRFN4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5140294
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: GEGTLESEPA VQVTEVTATS GLVSWGPGRP ADPVWMFQIQ YNSSEDETLI YRIVPASSHH FLLKHLVPGA DYDLC.
LRFN4 is one of a family of five transmembrane glycoproteins that are highly expressed in neuronal tissues. LRFN proteins share leucine-rich repeat (LRR)-immunoglobulin-like (Ig)-fibronectin type III (Fn)-transmembrane domain structure with other members of the LRR-Ig-Fn protein superfamily such as the Slitrk family of proteins. Expression of LRFN1, -3, and -4 mRNA was detected in embryonic neuronal cells, while Lrfn2 and Lrfn5 expression was primarily restricted to more mature cells. LRFN1, -2, and -4 bound to PDZ domains of postsynaptic PSD95, re-distributing PSD95 to the cell periphery. It has been suggested that the Lrfn proteins play a role in the developing and/or mature vertebrate nervous system. At least two isoforms of LRFN4 are known to exist.
Specifications
LRFN4 | |
Polyclonal | |
Unconjugated | |
LRFN4 | |
BC023156; fibronectin type III, immunoglobulin and leucine rich repeat domains 6; FIGLER6; leucine rich repeat and fibronectin type III domain containing 4; leucine-rich repeat and fibronectin type-III domain-containing protein 4; Lrfn4; Salm3; SALM3.; synaptic adhesion-like molecule 3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
78999 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6PJG9 | |
LRFN4 | |
Recombinant protein corresponding to Human LRFN4. Recombinant protein control fragment (Product #RP-109634). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction