Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRFN5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551529
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551529 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551529 Supplier Invitrogen™ Supplier No. PA551529
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PEGKLISNAT RSLVYDNGTL DILITTVKDT GAFTCIASNP AGEATQIVDL HIIKLPHLLN STNHIHEPDP GSSDISTSTK SGSNTSSSNG DTKLSQDKIV VAEATSSTAL LKFNFQRNIP GIRMFQIQYN GTYDDTLVYR MIPP Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 96%.

LRFN5 is one of a family of five transmembrane glycoproteins that are highly expressed in neuronal tissues. LRFN proteins share leucine-rich repeat (LRR)-immunoglobulin-like (Ig)-fibronectin type III (Fn)-transmembrane domain structure with other members of the LRR-Ig-Fn protein superfamily such as the Slitrk family of proteins. Expression of LRFN1, -3, and -4 mRNA was detected in embryonic neuronal cells, while LRFN2 and LRFN5 expression was primarily restricted to more mature cells. LRFN1, -2, and -4 bound to PDZ domains of postsynaptic PSD95, re-distributing PSD95 to the cell periphery. It has been suggested that the Lrfn proteins play a role in the developing and/or mature vertebrate nervous system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRFN5
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LRFN5
Gene Accession No. Q96NI6
Gene Alias AI427653; AI604817; C130061B21; C14orf146; fibronectin type III, immunoglobulin and leucine rich repeat domains 8; FIGLER8; Kiaa4208; leucine rich repeat and fibronectin type III domain containing 5; leucine-rich repeat and fibronectin type-III domain-containing protein 5; LRFN5; mKIAA4208; SALM5
Gene Symbols LRFN5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LRFN5. Recombinant protein control fragment (Product #RP-88982).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 145581
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.