Learn More
Invitrogen™ LRFN5 Polyclonal Antibody

Description
Immunogen sequence: PLITRHTHEM RVLEGQRATL RCKARGDPEP AIHWISPEGK LISNATRSLV For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 77%.
Specifications
Specifications
| Antigen | LRFN5 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | LRFN5 |
| Gene Accession No. | Q96NI6 |
| Gene Alias | AI427653; AI604817; C130061B21; C14orf146; fibronectin type III, immunoglobulin and leucine rich repeat domains 8; FIGLER8; Kiaa4208; leucine rich repeat and fibronectin type III domain containing 5; leucine-rich repeat and fibronectin type-III domain-containing protein 5; LRFN5; mKIAA4208; SALM5 |
| Gene Symbols | LRFN5 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.