Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRFN5 Rabbit anti-Human, Alexa Fluor 647, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306054AF647
Description
LRFN5 Polyclonal specifically detects LRFN5 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
LRFN5 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
C14orf146, chromosome 14 open reading frame 146, DKFZp686G0210, fibronectin type III, immunoglobulin and leucine rich repeat domains 8, FIGLER8, leucine rich repeat and fibronectin type III domain containing 5, leucine-rich repeat and fibronectin type-III domain-containing protein 5, SALM5FLJ30803 | |
Produced in rabbits immunized with E. coli-derived Human LRFN5 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
145581 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction