Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRG1 Antibody (2F5.A2), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NBP151830
Description
LRG1 Monoclonal specifically detects LRG1 in Human samples. It is validated for Immunoassay.Specifications
LRG1 | |
Monoclonal | |
0.1 mg/mL | |
Immunoassay 1 ug/ml | |
1300008B03Rik, 2310031E04Rik, HMFT1766, leucine-rich alpha-2-glycoprotein, leucine-rich alpha-2-glycoprotein 1, LRGleucine rich alpha 2 glycoprotein | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunoassay | |
2F5.A2 | |
Unconjugated | |
P02750 | |
LRG1 | |
Human LRG, covalently coupled to hemocyanin using glutaraldehyde | |
0.05 mg | |
Cancer | |
116844 | |
Store at 4°C. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction