Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRG1 Antibody (7A1), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
| Antigen | LRG1 |
|---|---|
| Clone | 7A1 |
| Dilution | Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200 |
| Applications | Western Blot, ELISA, Immunohistochemistry |
| Classification | Monoclonal |
Description
LRG1 Monoclonal antibody specifically detects LRG1 in Human samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
| LRG1 | |
| Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 1300008B03Rik, 2310031E04Rik, HMFT1766, leucine-rich alpha-2-glycoprotein, leucine-rich alpha-2-glycoprotein 1, LRGleucine rich alpha 2 glycoprotein | |
| A synthesized peptide derived from Human LRG1 [UniProt P02750] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 7A1 | |
| Western Blot, ELISA, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Cancer, Phospho Specific | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 116844 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title