Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRH-1/NR5A2 Antibody (OTI3H8), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272521AF532
Description
LRH-1/NR5A2 Monoclonal specifically detects LRH-1/NR5A2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
LRH-1/NR5A2 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
NR5A2 | |
Recombinant protein fragment corresponding to amino acids 1-303 of human LRH-1/NR5A2 (NP_995582) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI3H8 | |
Western Blot, Immunohistochemistry | |
Alpha-1-fetoprotein transcription factor, B1-binding factor, b1-binding factor, hepatocyte transcription factor which activates enhancer IIof hepatitis B virus, B1F, B1F2, fetoprotein-alpha 1 (AFP) transcription factor, FTFLRH1, FTZ-F1, FTZ-F1beta, HB1F, hB1F-2, hB1FCYP7A promoter-binding factor, Hepatocytic transcription factor, Liver receptor homolog 1, liver receptor homolog-1, LRH-1CPF, nuclear receptor NR5A2, nuclear receptor subfamily 5 group A member 2, nuclear receptor subfamily 5, group A, member 2 | |
Mouse | |
Affinity Purified | |
Cancer, Chromatin Research, GPCR, Neuroscience, Stem Cell Markers | |
2494.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction