Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRH-1/NR5A2 Antibody (SR2100), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | LRH-1/NR5A2 |
---|---|
Clone | SR2100 |
Dilution | Western Blot 1:500-1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
LRH-1/NR5A2 Monoclonal antibody specifically detects LRH-1/NR5A2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
LRH-1/NR5A2 | |
Western Blot 1:500-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
Alpha-1-fetoprotein transcription factor, B1-binding factor, b1-binding factor, hepatocyte transcription factor which activates enhancer IIof hepatitis B virus, B1F, B1F2, fetoprotein-alpha 1 (AFP) transcription factor, FTFLRH1, FTZ-F1, FTZ-F1beta, HB1F, hB1F-2, hB1FCYP7A promoter-binding factor, Hepatocytic transcription factor, Liver receptor homolog 1, liver receptor homolog-1, LRH-1CPF, nuclear receptor NR5A2, nuclear receptor subfamily 5 group A member 2, nuclear receptor subfamily 5, group A, member 2 | |
A synthesized peptide derived from human LRH-1/NR5A2 (Uniprot #: O00482) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR2100 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cancer, Chromatin Research, Neuroscience, Stem Cell Markers | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
2494 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title