Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRIG1 Antibody (789211), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB7498F
Description
LRIG1 Monoclonal antibody specifically detects LRIG1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOFSpecifications
LRIG1 | |
Monoclonal | |
FITC | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF | |
789211 | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF-ready | |
D6Bwg0781e, Img, LIG-1 | |
HEK293 human embryonic kidney cell line transfected with human LRIG1, Ala35-Ser779, Accession # Q96JA1 | |
0.1 mL | |
Cancer | |
26018 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction