Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRIG1 Mouse anti-Human, Clone: 2D1.1, MilliporeSigma™

Catalog No. MABS1816MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS1816MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS1816MI Supplier MilliporeSigma Supplier No. MABS1816
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-LRIG1 Antibody, clone 2D1.1 is a mouse monoclonal antibody that targets human Leucine-rich repeats and immunoglobulin-like domains protein 1 (LRIG1) and has been tested for use in Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRIG1
Applications Western Blot
Classification Monoclonal
Clone 2D1.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols LRIG1,LIG1
Host Species Mouse
Immunogen GST/His-tagged recombinant fragment corresponding to 134 amino acids from the C-terminal end of human Leucine-rich repeats and immunoglobulin-like domains protein 1 (LRIG1).
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_056356
Test Specificity Clone 2D1.1 detects Leucine-rich repeats and immunoglobulin-like domains protein 1 (LIG-1) in human. It targets an epitope within 134 amino acids from the C-terminal end.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.