Learn More
Invitrogen™ LRIG1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA595565
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Caco-2 whole cell. IHC: human mammary cancer tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
LRIG1 protein Q96JA1 is coded by a gene on a chromosome band 3p14.3, this region is known to be deleted in various human cancers. LRIG1 is considered to be a tumor suppressor gene in humans. LRIG1 is an integral cell-surface membrane protein that is expressed by specific cells in various human tissues and that its 143-kDa form might be cleaved into 111-kDa and 32-kDa fragments. The LRIG1 protein may inhibit the growth of tumors of glial cells and the down-regulation of the LRIG1 gene may be involved in the development and progression of the tumor.
Specifications
LRIG1 | |
Polyclonal | |
Unconjugated | |
Lrig1 | |
D6Bwg0781e; DKFZp586O1624; Img; integral membrane glycoprotein; leucine rich repeats and immunoglobulin like domains 1; leucine-rich repeat protein LRIG1; leucine-rich repeats and immunoglobulin like domains 1; leucine-rich repeats and immunoglobulin-like domains 1; leucine-rich repeats and immunoglobulin-like domains protein 1; LIG1; LIG-1; LRIG1; ortholog of mouse integral membrane glycoprotein LIG-1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
16206, 26018, 312574 | |
-20°C | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
500 μg/mL | |
PBS with 4mg trehalose and 0.05mg sodium azide | |
P70193, Q96JA1 | |
Lrig1 | |
A synthetic peptide corresponding to a sequence of mouse LRIG1 (AKRAFSGLESLEHLNLGENAIRSVQFDAFAKMKNLKELYI). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.