Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRIG1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984217
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4217 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4217 Supplier Bioss Supplier No. BS1844R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LRIG1 protein Q96JA1 is coded by a gene on a chromosome band 3p14.3, this region is known to be deleted in various human cancers. LRIG1 is considered to be a tumor suppressor gene in humans. LRIG1 is an integral cell-surface membrane protein that is expressed by specific cells in various human tissues and that its 143-kDa form might be cleaved into 111-kDa and 32-kDa fragments. The LRIG1 protein may inhibit the growth of tumors of glial cells and the down-regulation of the LRIG1 gene may be involved in the development and progression of the tumor.

Specifications

Antigen LRIG1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Lrig1
Gene Accession No. Q96JA1
Gene Alias D6Bwg0781e; DKFZp586O1624; Img; integral membrane glycoprotein; leucine rich repeats and immunoglobulin like domains 1; leucine-rich repeat protein LRIG1; leucine-rich repeats and immunoglobulin like domains 1; leucine-rich repeats and immunoglobulin-like domains 1; Leucine-rich repeats and immunoglobulin-like domains protein 1; LIG1; LIG-1; LRIG1; ortholog of mouse integral membrane glycoprotein LIG-1
Gene Symbols Lrig1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human LRIG1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26018
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.