Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRIG2 Antibody (990310), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB1941A
Description
LRIG2 Monoclonal antibody specifically detects LRIG2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
LRIG2 | |
Monoclonal | |
APC | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
9860 | |
Store at 4°C in the dark. | |
IgG1 |
Flow Cytometry, Immunocytochemistry, CyTOF | |
990310 | |
Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
KIAA0806, leucine-rich repeats and immunoglobulin-like domains 2, leucine-rich repeats and immunoglobulin-like domains protein 2, LIG2 | |
Chinese hamster ovary cell line CHO-derived recombinant human LRIG-2, Gly41-Thr805, Accession # O94898 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction