Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LRIG3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552737
Description
Immunogen sequence: NLCLNKSSLD FSANPEPASV ASSNSFMGTF GKALRRPHLD AYSSFGQPSD CQPRAFYLKA HSSPDLDSGS EEDGKERTDF QEENHICTFK QTLENYRTPN Highest antigen sequence identity to the following orthologs: Mouse - 69%, Rat - 68%.
May play a role in craniofacial and inner ear morphogenesis during embryonic development. May act within the otic vesicle epithelium to control formation of the lateral semicircular canal in the inner ear, possibly by restricting the expression of NTN1.
Specifications
LRIG3 | |
Polyclonal | |
Unconjugated | |
LRIG3 | |
leucine rich repeats and immunoglobulin like domains 3; leucine-rich repeats and immunoglobulin like domains 3; leucine-rich repeats and immunoglobulin-like domains 3; leucine-rich repeats and immunoglobulin-like domains protein 3; LIG3; LIG-3; LRIG3; RGD1561255; UNQ287/PRO326/PRO335 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
121227 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6UXM1 | |
LRIG3 | |
Recombinant protein corresponding to Human LRIG3. Recombinant protein control fragment (Product #RP-90294). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction