Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRIT3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553064
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553064 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553064 Supplier Invitrogen™ Supplier No. PA553064
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NPWFCDCHIS KMIELSKVVD PAIVLLDPLM TCSEPERLTG ILFQRAELEH CLKPSVMTSA TKIMSALGSN VLLRCDATGF PTPQITWTRS DSSPVNYTVI QESPEEGVRW SIMSLTGISS KDAGDYKCKA KNLAGMSEAV V Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 83%.

This gene encodes a protein that has a fibronectin type III domain and a C-terminal transmembrane domain, as well as a leucine-rich repeat domain and immunoglobulin-like domain near the N-terminus. The encoded protein may regulate fibroblast growth factor receptors and affect the modification of these receptors, which are glycosylated differently in the Golgi and endoplasmic reticulum. Mutations in this gene are associated with congenital stationary night blindness, type 1F.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRIT3
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LRIT3
Gene Accession No. Q3SXY7
Gene Alias CSNB1F; fibronectin type III, immunoglobulin and leucine rich repeat domains 4; FIGLER4; leucine rich repeat, Ig-like and transmembrane domains 3; leucine-rich repeat, Ig-like and transmembrane domains 3; leucine-rich repeat, immunoglobulin-like and transmembrane domains 3; leucine-rich repeat, immunoglobulin-like domain and transmembrane domain-containing protein 3; LRIT3
Gene Symbols LRIT3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LRIT3. Recombinant protein control fragment (Product #RP-90534).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 345193
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.