Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Lrmp Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553824
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553824 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553824 Supplier Invitrogen™ Supplier No. PA553824
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RSASLNSKPS SLRRVTIASL PRNIGNAGMV AGMENNDRFS RRSSSWRILG SKQSEHRPSL PRFISTYSWA DAEEEKCELK TKDDSEPSGE ETVERTRKPS LSEKKNNPSK WDVSSVYD Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 74%.

Plays a role in the delivery of peptides to major histocompatibility complex (MHC) class I molecules; this occurs in a transporter associated with antigen processing (TAP)-independent manner. May play a role in taste signal transduction via ITPR3. May play a role during fertilization in pronucleus congression and fusion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lrmp
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LRMP
Gene Accession No. Q12912
Gene Alias D6Int3; D6Int4; D6Int5; D6Int7; D6Int8; JAW1; LRMP; lymphoid restricted membrane protein; lymphoid-restricted membrane protein; Processed lymphoid-restricted membrane protein; Protein Jaw1
Gene Symbols LRMP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Lrmp. Recombinant protein control fragment (Product #RP-102121).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4033
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.