Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRP-1B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP27656625UL
Description
LRP-1B Polyclonal specifically detects LRP-1B in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
LRP-1B | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence 1-4 μg/mL | |
EC 1.1.1.94, EC 3.4.21.9, low density lipoprotein receptor related protein-deleted in tumor, low density lipoprotein receptor-related protein 1B, low density lipoprotein-related protein 1B (deleted in tumors), Low-density lipoprotein receptor-related protein-deleted in tumor, LRP-1B, LRP-deleted in tumors, LRPDIT, LRP-DITlow-density lipoprotein receptor-related protein 1B | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide | |
LRP1B | |
This antibody was developed against Recombinant Protein corresponding to amino acids: IANDTDILGFIYPFNYSGDHQQISHIEHNSRITGMDVYYQRDMIIWSTQFNPGGIFYKRIHGREKRQANSGLICPEF | |
25 μL | |
Cancer | |
53353.0 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction