Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRP-4 Antibody (S207-27), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222416B
Description
LRP-4 Monoclonal specifically detects LRP-4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
LRP-4 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
LRP4 | |
Fusion protein amino acids 26- 350 (extracellular N-terminus) of mouse LTP4 | |
0.1 mL | |
Primary | |
Detects the extracellular domain of LRP4 approx 215kDa. Degradation product can be seen at approx 150kDa. | |
Store at 4C in the dark. | |
IgG2a |
Western Blot, Immunocytochemistry, Immunofluorescence | |
S207-27 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
CLSS, KIAA0816, low density lipoprotein receptor-related protein 4, low-density lipoprotein receptor-related protein 4, LRP-4, MEGF7LRP10, Multiple epidermal growth factor-like domains 7 | |
Mouse | |
Protein G purified | |
Stem Cell Markers | |
4038 | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction