Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRP-4 Mouse anti-Human, Alexa Fluor 750, Clone: 814334, R&D Systems™

Catalog No. FA59481S100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FA59481S100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FA59481S100 Supplier R&D Systems Supplier No. FAB59481S100UG
This item is not returnable. View return policy

Mouse MonoclonalAntibody

Specifications

Antigen LRP-4
Applications Flow Cytometry
Classification Monoclonal
Clone 814334
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 750
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. O75096
Gene Alias CLSS, KIAA0816, low density lipoprotein receptor-related protein 4, low-density lipoprotein receptor-related protein 4, LRP10, LRP4, LRP-4, MEGF7LRP10, Multiple epidermal growth factor-like domains 7
Gene Symbols LRP4
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human LRP-4, Ser21-Ser1725 (predicted), Accession # O75096
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4038
Test Specificity Detects human LRP-4 in ELISAs.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 °C. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.