Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRP-5 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB1001886
Description
LRP-5 Polyclonal specifically detects LRP-5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.Specifications
LRP-5 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
BMND1OPTA1, EVR1, EVR4, HBM, low density lipoprotein receptor-related protein 5, low density lipoprotein receptor-related protein 7, low-density lipoprotein receptor-related protein 5, LR3VBCH2, LRP-5, LRP7exudative vitreoretinopathy 1, OPPG, OPS, osteoporosis pseudoglioma syndrome | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
Human. Expected from sequence similarity: Mouse and Rat. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
0.5 mg/mL | |
Western Blot 1-3 ug/ml, Immunohistochemistry 2-4 ug/ml, Immunohistochemistry-Paraffin 2-4 ug/ml, Peptide ELISA detection limit 1:16000 | |
O75197 | |
LRP5 | |
Peptide with sequence C-ERVEKTTGDKRT corresponding to internal region according to NP_002326.2. | |
0.1 mg | |
Cell Cycle and Replication, Stem Cell Signaling Pathway, Vision, Wnt Signaling Pathway | |
4041 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction