Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRP-5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP237868
Description
LRP-5 Polyclonal specifically detects LRP-5 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
LRP-5 | |
Polyclonal | |
Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
O75197 | |
LRP5 | |
This antibody was developed against a recombinant protein corresponding to amino acids: VVCQRYAGANGPFPHEYVSGTPHVPLNFIAPGGSQHGPFTGIACGKSMMSSVSLMGGRGGVPLYDRNHVTGASSSSSSSTKATLYPPILNPPPSPATDPSLYNMDMFYSSNIPATARPYRPYII | |
0.1 mL | |
Cell Cycle and Replication, Stem Cell Signaling Pathway, Vision, Wnt Signaling Pathway | |
4041 | |
Human | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
BMND1OPTA1, EVR1, EVR4, HBM, low density lipoprotein receptor-related protein 5, low density lipoprotein receptor-related protein 7, low-density lipoprotein receptor-related protein 5, LR3VBCH2, LRP-5, LRP7exudative vitreoretinopathy 1, OPPG, OPS, osteoporosis pseudoglioma syndrome | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction