Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRP-6 Antibody (255302) [DyLight 594], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB1505M
Description
LRP-6 Monoclonal antibody specifically detects LRP-6 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
LRP-6 | |
Monoclonal | |
DyLight 594 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunocytochemistry, CyTOF | |
255302 | |
Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
ADCAD2, FLJ90062, FLJ90421, low density lipoprotein receptor-related protein 6, low-density lipoprotein receptor-related protein 6, LRP-6 | |
Mouse myeloma cell line NS0-derived recombinant human LRP-6, Ala20-Pro1368, Accession # O75581 | |
0.1 mL | |
Stem Cell Signaling Pathway | |
4040 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction