Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRP-6 Antibody (SR1271), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | LRP-6 |
---|---|
Clone | SR1271 |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
Applications | Western Blot, Flow Cytometry, Immunofluorescence |
Classification | Monoclonal |
Description
LRP-6 Monoclonal antibody specifically detects LRP-6 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ImmunofluorescenceSpecifications
LRP-6 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
ADCAD2, FLJ90062, FLJ90421, low density lipoprotein receptor-related protein 6, low-density lipoprotein receptor-related protein 6, LRP-6 | |
A synthesized peptide derived from human LRP-6 (Uniprot #: O75581) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1271 | |
Western Blot, Flow Cytometry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Phospho Specific, Stem Cell Signaling Pathway | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
4040 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title