Learn More
Invitrogen™ LRP1 Monoclonal Antibody (Alpha 2MR alpha-2), PE

Description
A2Mr alpha-2 recognizes human CD91, also known as Prolow-density lipoprotein receptor-related protein 1, Alpha-2-macroglobulin receptor or apolipoprotein E receptor. CD91 is a 4525 amino acid protein post translationally cleaved into 3 subunits, a 85 kDa type I transmembrane carboxyl chain (LRP85) non-covalently bound to a 515 kDa extracellular N-terminal subunit (LRP515)containing multiple EGF-like and LDL-receptor Class A and Class B domains. A2Mr alpha-2 has been used extensively for the detection of CD91 by flow cytometry and immunohistochemistry.
Specifications
Specifications
| Antigen | LRP1 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | Alpha 2MR alpha-2 |
| Concentration | Conc. Not Determined |
| Conjugate | PE |
| Formulation | PBS with 1% BSA, 5% sucrose and 0.09% sodium azide |
| Gene | LRP1 |
| Gene Accession No. | Q07954 |
| Gene Alias | A2MR; AI316852; alpha 2-macroglobulin receptor; alpha-2-macroglobulin receptor; APOER; Apolipoprotein E receptor; APR; b2b1554Clo; CD91; FLJ16451; IGFBP3R; LDL receptor related protein 1; lipoprotein receptor-related protein; low density lipoprotein receptor-related protein 1; low density lipoprotein-related protein 1 (alpha-2-macroglobulin receptor); Low-density lipoprotein receptor-related protein 1 515 kDa subunit; Low-density lipoprotein receptor-related protein 1 85 kDa subunit; Low-density lipoprotein receptor-related protein 1 intracellular domain; Lrp; Lrp1; LRP-1; LRP1A; LRP-515; LRP-85; LRPICD; MGC88725; Prolow-density lipoprotein receptor-related protein 1; TbetaR-V/LRP-1/IGFBP-3 receptor; TGFBR5; type V tgf-beta receptor |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.